Anti-DNase I Rabbit Polyclonal Antibody

Leverandør: Abcam
AB113241-50
ABCAAB113241-50EA 12200 NOK
ABCAAB113241-50
Anti-DNase I Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: DNase I
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR