Anti-mSin3A Rabbit Polyclonal Antibody

Leverandør: Abcam
AB3479-100
ABCAAB3479-100EA 12550 NOK
ABCAAB3479-100
Anti-mSin3A Rabbit Polyclonal Antibody
Antistoffer
NB:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of

More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol MSIN3A
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human,Mouse
Vert Rabbit
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
Immunutfelling Yes
ImmunoFluorescens Yes
ChIP Yes
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR