Anti-JAK3 Rabbit Monoclonal Antibody [clone: EP909Y]

Leverandør: Abcam
AB45141-100
ABCAAB45141-100EA 12075 NOK
ABCAAB45141-100
Anti-JAK3 Rabbit Monoclonal Antibody [clone: EP909Y]
Antistoffer
Type: Primary
Antigen: JAK3 (Janus kinase 3)
Clonality: monoclonal
Clone: EP909Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR