Anti-JAK3 Rabbit Polyclonal Antibody

Leverandør: Abcam
AB61102-100
ABCAAB61102-100EA 12150 NOK
ABCAAB61102-100
Anti-JAK3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: JAK3 (Janus kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol JAK3
Antigen-navn Janus kinase 3
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Isotype IgG
ELISA Yes
ImmunoKjemi Yes
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR