Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: Abcam
AB6266-500
ABCAAB6266-500EA 6250 NOK
ABCAAB6266-500
Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
NB:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of

More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol IgG
Environmentally Preferable
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering FITC (Fluorescein Isothiocyanate)
Reaktivitet
Vert Goat
Isotype IgG
ELISA Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Flowcytometri Yes
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR