Anti-Fluorescein Goat Polyclonal Antibody (TRITC (Rhodamine))

Leverandør: Abcam
AB7253-100
ABCAAB7253-100EA 11600 NOK
ABCAAB7253-100
Anti-Fluorescein Goat Polyclonal Antibody (TRITC (Rhodamine))
Antistoffer
Type: Primary
Antigen: Fluorescein (fluorescein)
Clonality: polyclonal
Clone:
Conjugation: TRITC (Rhodamine)
Epitope:
Host: Goat
Isotype: none
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR