Anti-Spermidine Rabbit Polyclonal Antibody

Leverandør: Abcam
AB7318-200
ABCAAB7318-200EA 11675 NOK
ABCAAB7318-200
Anti-Spermidine Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Spermidine
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR