Anti-Semaphorin 4D Mouse Monoclonal Antibody [clone: 133-1C6]

Leverandør: Abcam
AB95612-100
ABCAAB95612-100EA 8050 NOK
ABCAAB95612-100
Anti-Semaphorin 4D Mouse Monoclonal Antibody [clone: 133-1C6]
Antistoffer
NB:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of

More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol SEMA4D
Antigen-navn Semaphorin 4D
Environmentally Preferable
Antistofftype Primary
Klonalitet Monoclonal
Konjugering Unconjugated
Klon 133-1C6
Reaktivitet Human
Vert Mouse
Isotype IgM
ELISA Yes
Immunutfelling Yes
Flowcytometri Yes
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR