Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: Abcam
AB97199-1
ABCAAB97199-1EA 5700 NOK
ABCAAB97199-1
Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR