Anti-EPHB4 Mouse Monoclonal Antibody

Leverandør: Abgent
AM7625A
ABGEAM7625AEA 4150 NOK
ABGEAM7625A
Anti-EPHB4 Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: EPHB4 (EPH receptor B4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR