Anti-MICALL1 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP10498A
ABGEAP10498AEA 4150 NOK
ABGEAP10498A
Anti-MICALL1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: MICALL1 (MICAL-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR