Anti-BCO2 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP11050A
ABGEAP11050AEA 4150 NOK
ABGEAP11050A
Anti-BCO2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: BCO2 (beta-carotene oxygenase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR