Anti-ZNF146 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP12388A
ABGEAP12388AEA 4150 NOK
ABGEAP12388A
Anti-ZNF146 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ZNF146 (zinc finger protein 146)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR