Anti-MATN1 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP12822B
ABGEAP12822BEA 4150 NOK
ABGEAP12822B
Anti-MATN1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: MATN1 (matrilin 1, cartilage matrix protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR