Anti-CTBP2 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP12902A
ABGEAP12902AEA 4150 NOK
ABGEAP12902A
Anti-CTBP2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CTBP2 (C-terminal binding protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR