Anti-ARFIP1 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP12992C
ABGEAP12992CEA 4150 NOK
ABGEAP12992C
Anti-ARFIP1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ARFIP1 (ADP-ribosylation factor interacting protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR