Anti-BCL2 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP1303A
ABGEAP1303AEA 4150 NOK
ABGEAP1303A
Anti-BCL2 Rabbit Polyclonal Antibody
Antistoffer
This apoptosis regulator Bcl antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 75-110 amino acids from human Bcl. Including factor-dependent lymphohematopoietic and neural cells this antibody suppresses apoptosis.

  • Controls mitochondrial membrane permeability
  • Inhibits caspase activity
  • Expressed in a variety of tissues

It prevents the release of cytochrome c from the mitochondria and/or by binding to the apoptosis-activating factor (APAF-1) and hence inhibits the caspase activity.

Type: Primary
Antigen: BCL2
More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol BCL2
Environmentally Preferable
Antigen-navn B-cell lymphoma 2
Antigen synonymer PPP1R50|Bcl-2
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human
Vert Rabbit
Isotype
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
Immunutfelling Yes
Epitop 50–150 aa
Kyssabsorpsjon No
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR