Anti-AIFM2 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP1355C
ABGEAP1355CEA 4150 NOK
ABGEAP1355C
Anti-AIFM2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: AIFM2 (apoptosis-inducing factor, mitochondrion-associated, 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR