Anti-MGMT Rabbit Polyclonal Antibody

Leverandør: Abgent
AP13848A
ABGEAP13848AEA 4150 NOK
ABGEAP13848A
Anti-MGMT Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: MGMT (O-6-methylguanine-DNA methyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR