Anti-PAK3 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP13931A
ABGEAP13931AEA 4150 NOK
ABGEAP13931A
Anti-PAK3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PAK3 (p21 protein (Cdc42/Rac)-activated kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR