Anti-PSMA5 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP1449C
ABGEAP1449CEA 4150 NOK
ABGEAP1449C
Anti-PSMA5 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PSMA5 (proteasome (prosome, macropain) subunit, alpha type, 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR