Anti-KEL Rabbit Polyclonal Antibody

Leverandør: Abgent
AP14875C
ABGEAP14875CEA 4150 NOK
ABGEAP14875C
Anti-KEL Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: KEL (Kell blood group, metallo-endopeptidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR