Anti-TLR4 rabbit polyclonal antibody

Leverandør: Abgent
AP1504A
ABGEAP1504AEA 4150 NOK
ABGEAP1504A
Anti-TLR4 rabbit polyclonal antibody
Antistoffer
Type: Primary
Antigen: TLR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal ( 39–55 aa)
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR