Anti-TAS2R31 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP16600B
ABGEAP16600BEA 4150 NOK
ABGEAP16600B
Anti-TAS2R31 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: TAS2R31
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR