Anti-FAIM Rabbit Polyclonal Antibody

Leverandør: Abgent
AP16625C
ABGEAP16625CEA 4150 NOK
ABGEAP16625C
Anti-FAIM Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: FAIM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR