Anti-PRODH2 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP16627C
ABGEAP16627CEA 4150 NOK
ABGEAP16627C
Anti-PRODH2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PRODH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR