Anti-NHLH2 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP16640C
ABGEAP16640CEA 4150 NOK
ABGEAP16640C
Anti-NHLH2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: NHLH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR