Anti-MPI Rabbit Polyclonal Antibody

Leverandør: Abgent
AP17091A
ABGEAP17091AEA 4150 NOK
ABGEAP17091A
Anti-MPI Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: MPI (mannose phosphate isomerase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol MPI
Antigen-navn mannose phosphate isomerase
Antigen synonymer PMI|PMI1|CDG1B
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human
Vert Rabbit
Gen-ID 4351
Western blot Yes
ELISA Yes
Flowcytometri Yes
Epitop N-Terminal
Immunogen This MPI antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 37-65 amino acids from the N-terminal region of human MPI.
Sekvens GTHPRGDAKILDNRC
Molekylvekt 46.656 kDa
Rensing Protein A & Peptide affinity purification
Konsentrasjon 0,5 mg/ml
Lagringsbuffer PBS with 0,09% (W/V) sodium azide, pH 7,4
Lagringstemp Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
Frakttemp. 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR