Anti-AP4M1 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP17446C
ABGEAP17446CEA 4150 NOK
ABGEAP17446C
Anti-AP4M1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: AP4M1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol AP4M1
Environmentally Preferable
Antigen-navn adaptor-related protein complex 4, mu 1 subunit
Antigen synonymer SPG50|CPSQ3|MU-4|MU-ARP2
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human
Vert Rabbit
Gen-ID 9179
Isotype
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
Flowcytometri Yes
Epitop internal
Immunogen This AP4M1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 223-251 amino acids from the Central region of human AP4M1.
Aminosyretall 223 to 251
Sekvens KSELRGYGPGIRVDC
Molekylvekt 49.977 kDa
Rensing Protein A & Peptide affinity purification
Konsentrasjon 0.42
Lagringsbuffer pH 7,4
Lagringstemp Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
Frakttemp. 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR