Anti-LRRC42 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP17990C
ABGEAP17990CEA 4150 NOK
ABGEAP17990C
Anti-LRRC42 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: LRRC42
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol LRRC42
Antigen-navn leucine rich repeat containing 42
Environmentally Preferable
Antigen synonymer dJ167A19.4
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human
Vert Rabbit
Gen-ID 115353
Isotype
Western blot Yes
ELISA Yes
Flowcytometri Yes
Epitop internal
Immunogen This LRRC42 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 44-71 amino acids from the Central region of human LRRC42.
Aminosyretall 44 to 71
Sekvens REDDTARKEKTDHC
Molekylvekt 48.571 kDa
Rensing Protein A & Peptide affinity purification
Konsentrasjon 1.090
Lagringsbuffer pH 7,4
Lagringstemp Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
Frakttemp. 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR