Anti-HEPHL1 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP18557A
ABGEAP18557AEA 4150 NOK
ABGEAP18557A
Anti-HEPHL1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: HEPHL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR