Anti-ARHGEF16 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP19783A
ABGEAP19783AEA 4150 NOK
ABGEAP19783A
Anti-ARHGEF16 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ARHGEF16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR