Anti-STAT3 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP19819B
ABGEAP19819BEA 4150 NOK
ABGEAP19819B
Anti-STAT3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: STAT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR