Anti-PABPC1 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP2920C
ABGEAP2920CEA 4150 NOK
ABGEAP2920C
Anti-PABPC1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PABPC1 (poly(A) binding protein, cytoplasmic 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR