Anti-CD80 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP5020B
ABGEAP5020BEA 4150 NOK
ABGEAP5020B
Anti-CD80 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CD80 (CD80 molecule)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR