Anti-TAP1 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP6252A
ABGEAP6252AEA 4150 NOK
ABGEAP6252A
Anti-TAP1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: TAP1 (transporter 1, ATP-binding cassette, sub-family B (MDR/TAP))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR