Anti-LATS1 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP7035A
ABGEAP7035AEA 4150 NOK
ABGEAP7035A
Anti-LATS1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: LATS1 (large tumor suppressor kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR