Anti-CKMT2 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP7072B
ABGEAP7072BEA 4150 NOK
ABGEAP7072B
Anti-CKMT2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CKMT2 (creatine kinase, mitochondrial 2 (sarcomeric))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR