Anti-RASGRF1 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP7582B
ABGEAP7582BEA 4150 NOK
ABGEAP7582B
Anti-RASGRF1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: RASGRF1 (Ras protein-specific guanine nucleotide-releasing factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR