Anti-MUSK Rabbit Polyclonal Antibody

Leverandør: Abgent
AP7664D
ABGEAP7664DEA 4150 NOK
ABGEAP7664D
Anti-MUSK Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: MUSK (muscle, skeletal, receptor tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR