Anti-ALDH4A1 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP7875C
ABGEAP7875CEA 4150 NOK
ABGEAP7875C
Anti-ALDH4A1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ALDH4A1 (aldehyde dehydrogenase 4 family, member A1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR