Anti-CYP2C8 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP7995A
ABGEAP7995AEA 4150 NOK
ABGEAP7995A
Anti-CYP2C8 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CYP2C8 (cytochrome P450, family 2, subfamily C, polypeptide 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR