Anti-C5 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP8577A
ABGEAP8577AEA 4150 NOK
ABGEAP8577A
Anti-C5 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: C5 (complement component 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR