Anti-HAO1 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP8605C
ABGEAP8605CEA 4150 NOK
ABGEAP8605C
Anti-HAO1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: HAO1 (hydroxyacid oxidase (glycolate oxidase) 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR