Anti-GLIS1 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP8878A
ABGEAP8878AEA 4150 NOK
ABGEAP8878A
Anti-GLIS1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: GLIS1 (GLIS family zinc finger 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR