Anti-PLCB2 Rabbit Polyclonal Antibody

Leverandør: Abgent
AP9253A
ABGEAP9253AEA 4150 NOK
ABGEAP9253A
Anti-PLCB2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PLCB2 (phospholipase C, beta 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR