Anti-JAK3 Mouse Monoclonal Antibody [clone: 3C2]

Leverandør: Abnova
H00003718-M01A
ABNOH00003718-M01AEA 4650 NOK
ABNOH00003718-M01A
Anti-JAK3 Mouse Monoclonal Antibody [clone: 3C2]
Antistoffer
Type: Primary
Antigen: JAK3
Clonality: Monoclonal
Clone: 3C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR