Anti-ZIC2 (Zic Family member 2) Mouse Monoclonal Antibody [clone: 3G5]

Leverandør: Abnova
H00007546-M02
ABNOH00007546-M02EA 4650 NOK
ABNOH00007546-M02
Anti-ZIC2 (Zic Family member 2) Mouse Monoclonal Antibody [clone: 3G5]
Antistoffer
Type: Primary
Antigen: ZIC2 (Zic Family member 2)
Clonality: Monoclonal
Clone: 3G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR