KLH IgG (Rat) ELISA kit
Leverandør: Abnova
KA6735.1KIT
ABNOKA6735.1KITEA 11700 NOK
ABNOKA6735.1KIT
KLH IgG (Rat) ELISA kit
Assay ELISAs
KLH IgG (Rat) ELISA Kit is for the quantitative measurement of rat anti-KLH IgG in serum or plasma samples.