KLH IgG (Human) ELISA kit
Leverandør: Abnova
KA6737.1KIT
ABNOKA6737.1KITEA 13350 NOK
ABNOKA6737.1KIT
KLH IgG (Human) ELISA kit
Assay ELISAs
KLH IgG (Human) ELISA Kit is for the quantitative measurement of human anti-KLH IgG in serum, plasma and other fluids.